This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
TTLL7
- Gene Name:
-
tubulin tyrosine ligase like 7
- Gene Aliases:
-
-
- Chromosome Location:
-
Chr.1: 83865021 - 83999340 on Build GRCh38
- UniGene ID:
-
Hs.445826
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
TTLL7 |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_024686.4 |
NP_078962.4 |
7, 8
|
1118
|
|
| XM_005271208.2 |
XP_005271265.1 |
6, 7
|
929
|
|
| XM_011542159.2 |
XP_011540461.1 |
7, 8
|
1308
|
|
| XM_017002347.1 |
XP_016857836.1 |
8, 9
|
1138
|
|
| XM_017002348.1 |
XP_016857837.1 |
7, 8
|
1308
|
|
| GenBank mRNA |
AK094174.1 |
- |
2, 3
|
720
|
|
| AK126792.1 |
- |
7, 8
|
1015
|
|
| AK128604.1 |
- |
4, 5
|
1097
|
|
| AY170843.1 |
- |
6, 7
|
892
|
|
| BC048970.1 |
- |
7, 8
|
1060
|
5295193 |
| BC060878.1 |
- |
7, 8
|
1100
|
5270163 |
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System