Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC7 SIRNA Assays ›
Gene Symbol:
CCDC7
Gene Name:
coiled-coil domain containing 7
Gene Aliases:
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
Chromosome Location:
Chr.10: 32446082 - 32882864 on Build GRCh38
UniGene ID:
Hs.585464
Species:
Human
siRNA ID
HSS128780
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321115.1
NP_001308044.1
38, 39
4289
XM_005252593.2
XP_005252650.1
19, 20
2076
XM_006717505.1
XP_006717568.1
36, 37
3784
XM_006717506.1
XP_006717569.1
35, 36
3676
XM_011519663.1
XP_011517965.1
38, 39
3997
XM_011519664.1
XP_011517966.1
38, 39
3997
XM_011519666.1
XP_011517968.1
37, 38
3913
XM_011519667.1
XP_011517969.1
37, 38
3913
XM_011519668.1
XP_011517970.1
37, 38
3907
XM_011519669.1
XP_011517971.1
37, 38
3895
XM_011519670.1
XP_011517972.1
37, 38
3889
XM_011519671.1
XP_011517973.1
37, 38
3889
XM_011519672.1
XP_011517974.1
37, 38
3886
XM_011519673.1
XP_011517975.1
37, 38
3883
XM_011519674.1
XP_011517976.1
36, 37
3805
XM_011519675.1
XP_011517977.1
36, 37
3787
XM_011519676.1
XP_011517978.1
36, 37
3775
XM_011519677.1
XP_011517979.1
37, 38
3775
XM_011519678.1
XP_011517980.1
36, 37
3772
XM_011519679.1
XP_011517981.1
35, 36
3466
XM_011519680.2
XP_011517982.1
33, 34
3132
XM_011519683.2
XP_011517985.1
27, 28
2806
XM_011519685.1
XP_011517987.1
30, 31
3014
XM_011519687.1
XP_011517989.1
20, 21
1921
XM_017016638.1
XP_016872127.1
38, 39
3952
XM_017016639.1
XP_016872128.1
37, 38
3886
XM_017016640.1
XP_016872129.1
36, 37
3805
XM_017016641.1
XP_016872130.1
33, 34
3426
XM_017016642.1
XP_016872131.1
33, 34
3499
XM_017016643.1
XP_016872132.1
33, 34
3116
XM_017016645.1
XP_016872134.1
32, 33
3040
XM_017016646.1
XP_016872135.1
29, 30
2927
XM_017016648.1
XP_016872137.1
25, 26
2648
XM_017016649.1
XP_016872138.1
25, 26
2588
XM_017016652.1
XP_016872141.1
22, 23
2051
XM_017016653.1
XP_016872142.1
20, 21
1961
GenBank mRNA
AK023093.1
-
17, 18
1890
AK097668.1
-
11, 12
1030
AK301824.1
-
16, 17
1472
BC125090.1
-
18, 19
1912
40118176
BC125091.1
-
16, 17
1499
40118177
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-p89b7:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0