Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077186.1 | NP_001070654.1 | 25 | 3203 | ||
NM_001145809.1 | NP_001139281.1 | 26 | 3302 | |||
NM_024729.3 | NP_079005.3 | 24 | 3179 | |||
XM_006723386.3 | XP_006723449.1 | 25 | 3264 | |||
XM_011527320.2 | XP_011525622.1 | 25 | 3281 | |||
XM_011527321.2 | XP_011525623.1 | 24 | 3257 | |||
XM_011527323.2 | XP_011525625.1 | 25 | 3271 | |||
GenBank mRNA | AB111886.1 | - | 15 | 1861 | ||
AB290169.1 | - | 26 | 3272 | |||
AY165122.1 | - | 25 | 3178 | |||
AY203926.1 | - | 3 | 244 |