Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLTM SIRNA Assays ›
Gene Symbol:
SLTM
Gene Name:
SAFB like transcription modulator
Gene Aliases:
Met
Chromosome Location:
Chr.15: 58879045 - 58934313 on Build GRCh38
UniGene ID:
Hs.512932
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS128902
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
459.00
Your Price
Online offer:
459.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLTM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001013843.2
NP_001013865.1
18, 19
2740
NM_024755.3
NP_079031.2
18, 19
2794
NR_135042.1
16, 17
2127
NR_135043.1
16, 17
2237
XM_006720686.3
XP_006720749.3
16, 17
3185
XM_006720690.1
XP_006720753.1
14, 15
1891
XM_011522022.1
XP_011520324.1
18, 19
3452
XM_011522023.1
XP_011520325.1
18, 19
3398
XM_011522024.1
XP_011520326.1
17, 18
3302
XM_011522025.1
XP_011520327.1
17, 18
3254
XM_011522026.2
XP_011520328.2
17, 18
3233
XM_011522027.1
XP_011520329.1
16, 17
3206
XM_011522028.1
XP_011520330.1
16, 17
3152
XM_011522029.2
XP_011520331.1
18, 19
2633
XM_011522031.2
XP_011520333.1
15, 16
2288
XM_017022576.1
XP_016878065.1
17, 18
2590
XM_017022577.1
XP_016878066.1
17, 18
3179
XM_017022578.1
XP_016878067.1
16, 17
3131
XM_017022579.1
XP_016878068.1
18, 19
2797
XM_017022580.1
XP_016878069.1
17, 18
2251
GenBank mRNA
AK023275.1
-
8, 9
1248
AK055195.1
-
16, 17
2030
AK291423.1
-
16, 17
2160
AL834297.1
-
4, 5
689
BC014944.1
-
11, 12
1597
BC046119.1
-
7, 8
1089
BC140851.1
-
18, 19
2761
9021719
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
nucleotide binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-4p4qf:80/100.66.72.111:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0