Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024756.2 | NP_079032.2 | 7 | 2963 | ||
XM_005270153.1 | XP_005270210.1 | 7 | 3002 | |||
XM_006717970.3 | XP_006718033.1 | 7 | 4942 | |||
GenBank mRNA | AK023527.1 | - | 7 | 2910 | ||
AK293822.1 | - | 6 | 2244 | |||
AK304727.1 | - | 7 | 2820 | |||
BC094744.1 | - | 7 | 2910 | 30343159 |