Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSPP1 SIRNA Assays ›
Gene Symbol:
CSPP1
Gene Name:
centrosome and spindle pole associated protein 1
Gene Aliases:
CSPP, JBTS21
Chromosome Location:
Chr.8: 67062417 - 67196614 on Build GRCh38
UniGene ID:
Hs.370147
Species:
Human
siRNA ID
HSS128988
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CSPP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291339.1
NP_001278268.1
9
1547
NM_024790.6
NP_079066.5
11
1394
XM_005251305.3
XP_005251362.2
14
1878
XM_006716474.2
XP_006716537.2
14
1878
XM_011517598.1
XP_011515900.1
15
1920
XM_011517599.1
XP_011515901.1
14
1896
XM_011517600.1
XP_011515902.1
13
1854
XM_011517601.1
XP_011515903.1
13
1815
XM_011517602.1
XP_011515904.1
12
1773
XM_011517603.1
XP_011515905.1
14
1603
XM_011517607.1
XP_011515909.1
13
1483
XM_011517608.1
XP_011515910.1
12
1480
XM_011517609.1
XP_011515911.1
11
1602
XM_017013847.1
XP_016869336.1
13
1779
XM_017013848.1
XP_016869337.1
12
1755
XM_017013849.1
XP_016869338.1
12
1716
XM_017013850.1
XP_016869339.1
11
1674
XM_017013851.1
XP_016869340.1
11
1438
XM_017013852.1
XP_016869341.1
11
1674
XM_017013853.1
XP_016869342.1
11
1438
XM_017013857.1
XP_016869346.1
7
957
XM_017013858.1
XP_016869347.1
6
858
XM_017013859.1
XP_016869348.1
1
91
GenBank mRNA
AJ583433.1
-
9
1547
AK096205.1
-
12
1448
AK127108.1
-
13
2851
AM156947.1
-
11
1395
BX649147.1
-
11
1395
Back To Top
More Information
Panther Classification:
Molecular Function -
centromere DNA-binding protein
Gene Ontology Categories:
Function(s)
positive regulation of cytokinesis
positive regulation of cell division
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-p89b7:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0