Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L2HGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024884.2 | NP_079160.1 | 5 | 714 | ||
XM_005268075.4 | XP_005268132.1 | 5 | 714 | |||
XM_011537166.2 | XP_011535468.1 | 6 | 821 | |||
XM_011537167.2 | XP_011535469.1 | 5 | 1099 | |||
XM_011537168.2 | XP_011535470.1 | 3 | 333 | |||
XM_017021655.1 | XP_016877144.1 | 5 | 845 | |||
XM_017021656.1 | XP_016877145.1 | 6 | 799 | |||
XM_017021657.1 | XP_016877146.1 | 6 | 726 | |||
XM_017021658.1 | XP_016877147.1 | 5 | 714 | |||
GenBank mRNA | AK022680.1 | - | 5 | 714 | ||
AK292627.1 | - | 5 | 714 | |||
AK309294.1 | - | 5 | 711 | |||
AY757363.1 | - | 5 | 649 | |||
BC006117.1 | - | 5 | 667 | 3504996 |