Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261392.1 | NP_001248321.1 | 11 | 981 | ||
NM_001261394.1 | NP_001248323.1 | 11 | 909 | |||
NM_024894.3 | NP_079170.2 | 12 | 1059 | |||
NR_048552.1 | 11 | 1013 | ||||
XM_011510398.2 | XP_011508700.1 | 11 | 926 | |||
XM_011510399.2 | XP_011508701.1 | 10 | 864 | |||
XM_011510400.2 | XP_011508702.1 | 12 | 1047 | |||
XM_011510402.2 | XP_011508704.1 | 12 | 1047 | |||
GenBank mRNA | AK024000.1 | - | 11 | 900 | ||
AK024137.1 | - | 11 | 894 | |||
AK290680.1 | - | 12 | 1023 | |||
AK297169.1 | - | 11 | 969 | |||
AK299617.1 | - | 11 | 1001 | |||
BC005125.2 | - | 12 | 1039 | 3841643 | ||
BG390065.1 | - | 7 | 513 |