Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024915.3 | NP_079191.2 | 11 | 1718 | ||
XM_011517305.2 | XP_011515607.1 | 11 | 1749 | |||
XM_011517306.2 | XP_011515608.1 | 11 | 1486 | |||
XM_011517307.2 | XP_011515609.1 | 11 | 1716 | |||
GenBank mRNA | AK023844.1 | - | 11 | 1400 | ||
AK292853.1 | - | 11 | 1400 | |||
BC069618.1 | - | 11 | 1400 | 7262389 | ||
BC069633.1 | - | 11 | 1460 | 7262401 | ||
BC069638.1 | - | 11 | 1400 | 7262413 | ||
BC129822.1 | - | 11 | 1400 | 40113571 | ||
BC129823.1 | - | 11 | 1400 | 40113576 |