Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008695.1 | NP_001008695.1 | 4 | 308 | ||
NM_030573.2 | NP_085050.2 | 3 | 482 | |||
GenBank mRNA | AK024609.1 | - | 1 | 1046 | ||
AK025906.1 | - | 1 | 814 | |||
AK315457.1 | - | 3 | 467 | |||
BC004346.1 | - | 3 | 453 | 3633743 | ||
BE264280.1 | - | 3 | 381 | |||
BI223500.1 | - | 3 | 425 | |||
BM790105.1 | - | 4 | 308 | |||
CR456343.1 | - | 3 | 300 |