Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR51E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030774.3 | NP_110401.1 | 2 | 1138 | ||
GenBank mRNA | AF369708.1 | - | 2 | 1044 | ||
AK314051.1 | - | 2 | 1138 | |||
AY033942.1 | - | 2 | 1135 | |||
BC020768.1 | - | 3 | 1011 | 4249775 |