Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NDEL1 SIRNA Assays ›
          Gene Symbol:
          NDEL1
          Gene Name:
          nudE neurodevelopment protein 1 like 1
          Gene Aliases:
          EOPA, MITAP1, NDE1L1, NDE2, NUDEL
          Chromosome Location:
          Chr.17: 8435852 - 8472744 on Build GRCh38
          UniGene ID:
          Hs.372123
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS129920
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NDEL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025579.2 NP_001020750.1 4 495
          NM_030808.4 NP_110435.1 4 495
          XM_011524013.1 XP_011522315.1 4 486
          XM_017025183.1 XP_016880672.1 5 675
          XM_017025184.1 XP_016880673.1 5 675
          XM_017025185.1 XP_016880674.1 4 544
          XM_017025186.1 XP_016880675.1 5 675
          XM_017025187.1 XP_016880676.1 4 544
          XM_017025188.1 XP_016880677.1 5 675
          GenBank mRNA AF182078.1 - 4 422
          AF217798.3 - 4 486
          AK056014.1 - 4 493
          AK299559.1 - 5 469
          AL832648.1 - 4 429
          AY004871.1 - 4 421
          BC026101.2 - 4 433 4820855
          BP201698.1 - 4 495
          CX166766.1 - 4 424
          DB176966.1 - 4 495
          DN993952.1 - 4 486

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_133320, XM_006246575, XM_006246573, XM_006246571, XM_006246572, and others.
          This siRNA also targets the following transcripts in Mouse: NM_023668, AF323918, AK146029, AF290472, BC046796, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of mitotic spindle orientation
          neuron migration
          inner cell mass cell proliferation
          proteolysis
          microtubule nucleation
          chromosome segregation
          sister chromatid cohesion
          mitotic centrosome separation
          retrograde axonal transport
          regulation of neuron projection development
          cell migration
          cerebral cortex radially oriented cell migration
          central nervous system neuron axonogenesis
          regulation of intracellular protein transport
          positive regulation of GTPase activity
          positive regulation of axon extension
          vesicle transport along microtubule
          positive regulation of axon regeneration
          nuclear envelope disassembly
          establishment of chromosome localization
          centrosome localization
          neurofilament cytoskeleton organization
          activation of GTPase activity
          neuron projection extension
          regulation of microtubule motor activity
          protein binding
          microtubule binding
          protein complex binding
          alpha-tubulin binding
          beta-tubulin binding
          oligopeptidase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0