Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FIP1L1 SIRNA Assays ›
Gene Symbol:
FIP1L1
Gene Name:
factor interacting with PAPOLA and CPSF1
Gene Aliases:
FIP1, Rhe, hFip1
Chromosome Location:
Chr.4: 53377572 - 53462611 on Build GRCh38
UniGene ID:
Hs.555109
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS129968
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FIP1L1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134937.1
NP_001128409.1
4, 5
466
NM_001134938.1
NP_001128410.1
4, 5
466
NM_030917.3
NP_112179.2
5, 6
511
XM_005265769.4
XP_005265826.1
5, 6
578
XM_005265773.4
XP_005265830.1
5, 6
580
XM_005265774.4
XP_005265831.1
5, 6
519
XM_005265778.4
XP_005265835.1
5, 6
563
XM_005265779.4
XP_005265836.1
5, 6
518
XM_005265781.4
XP_005265838.1
5, 6
572
XM_005265782.4
XP_005265839.1
5, 6
518
XM_017008662.1
XP_016864151.1
4, 5
483
XM_017008663.1
XP_016864152.1
5, 6
520
XM_017008664.1
XP_016864153.1
4, 5
536
XM_017008665.1
XP_016864154.1
5, 6
518
XM_017008666.1
XP_016864155.1
4, 5
496
XM_017008667.1
XP_016864156.1
4, 5
474
XM_017008668.1
XP_016864157.1
4, 5
547
XM_017008669.1
XP_016864158.1
4, 5
489
XM_017008670.1
XP_016864159.1
5, 6
518
XM_017008671.1
XP_016864160.1
5, 6
518
GenBank mRNA
AK295737.1
-
4, 5
481
AK300398.1
-
5, 6
321
AK308145.1
-
5, 6
321
AL136910.1
-
4, 5
447
AY366510.1
-
5, 6
468
BC017724.1
-
4, 5
448
3452247
BC024016.1
-
3, 4
1622
4826276
BC052959.1
-
3, 4
1617
4838737
BC110383.1
-
5, 6
504
5165430
DN990103.1
-
5, 6
520
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
mRNA export from nucleus
mRNA 3'-end processing
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-dsplp:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0