Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NIPA2 SIRNA Assays ›
Gene Symbol:
NIPA2
Gene Name:
non imprinted in Prader-Willi/Angelman syndrome 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 22838641 - 22868384 on Build GRCh38
UniGene ID:
Hs.591003
Species:
Human
siRNA ID
HSS129976
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NIPA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008860.2
NP_001008860.1
7
988
NM_001008892.2
NP_001008892.1
6
852
NM_001008894.2
NP_001008894.1
5
795
NM_001184888.1
NP_001171817.1
6
943
NM_001184889.1
NP_001171818.1
10
1335
NM_030922.6
NP_112184.4
8
1110
XM_005272546.3
XP_005272603.1
8
1343
XM_005272547.4
XP_005272604.1
8
1127
XM_005272548.3
XP_005272605.1
7
957
XM_005272550.3
XP_005272607.1
7
795
XM_005272552.3
XP_005272609.1
6
1057
XM_005272553.4
XP_005272610.1
6
861
XM_006720364.2
XP_006720427.1
9
1237
XM_006720365.2
XP_006720428.1
8
1082
XM_006720366.3
XP_006720429.1
7
948
XM_006720367.1
XP_006720430.1
7
1286
XM_011543877.2
XP_011542179.1
9
1470
XM_011543878.2
XP_011542180.1
9
1637
XM_011543879.2
XP_011542181.1
9
1208
XM_011543880.2
XP_011542182.1
7
1221
XM_017022645.1
XP_016878134.1
9
1531
XM_017022646.1
XP_016878135.1
9
1298
XM_017022647.1
XP_016878136.1
9
1246
XM_017022648.1
XP_016878137.1
9
1307
XM_017022649.1
XP_016878138.1
9
1441
XM_017022650.1
XP_016878139.1
9
1404
XM_017022651.1
XP_016878140.1
10
1825
XM_017022652.1
XP_016878141.1
9
1217
XM_017022653.1
XP_016878142.1
10
1531
XM_017022654.1
XP_016878143.1
8
1055
XM_017022655.1
XP_016878144.1
7
1053
XM_017022656.1
XP_016878145.1
7
1063
XM_017022657.1
XP_016878146.1
8
1180
XM_017022658.1
XP_016878147.1
6
901
XM_017022659.1
XP_016878148.1
8
1151
XM_017022660.1
XP_016878149.1
6
1164
XM_017022661.1
XP_016878150.1
6
931
XM_017022662.1
XP_016878151.1
5
1000
XM_017022663.1
XP_016878152.1
5
805
GenBank mRNA
AK096305.1
-
7
969
AK300843.1
-
5
738
AK313584.1
-
6
927
AY732242.1
-
10
1256
BC000957.3
-
3
281
BC011775.2
-
6
764
3640970
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
magnesium ion transport
magnesium ion transmembrane transport
magnesium ion transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-6dgdk:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0