Search
Search

TTLL2| Gene Symbol: TTLL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031949.4 | NP_114155.4 | 3 | 1195 | ||
| XM_006715572.3 | XP_006715635.1 | 2 | 990 | |||
| GenBank mRNA | AK093039.1 | - | 3 | 1195 | ||
| AK098527.1 | - | 1 | 636 | |||
| AK225984.1 | - | 3 | 1195 | |||
| AK301755.1 | - | 4 | 1052 | |||
| AK314450.1 | - | 3 | 1172 | |||
| AY026506.1 | - | 3 | 1167 | |||
| BC030650.1 | - | 3 | 1185 | 5269216 | ||
| BC047411.1 | - | 3 | 1173 | 5296230 | ||