Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSSK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032037.3 | NP_114426.1 | 1 | 328 | ||
GenBank mRNA | AF329483.1 | - | 1 | 217 | ||
AF348077.1 | - | 1 | 353 | |||
AK058166.1 | - | 1 | 216 | |||
AK172841.1 | - | 1 | 697 | |||
AK313767.1 | - | 1 | 216 | |||
BC014611.2 | - | 1 | 216 | 4826305 |