Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP13A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032279.3 | NP_115655.2 | 19 | 2581 | ||
XM_011513232.2 | XP_011511534.1 | 19 | 2328 | |||
XM_017007318.1 | XP_016862807.1 | 19 | 2217 | |||
XM_017007319.1 | XP_016862808.1 | 19 | 2328 | |||
GenBank mRNA | AK095277.1 | - | 19 | 2366 | ||
AY823162.1 | - | 19 | 2258 | |||
BC101496.1 | - | 19 | 2352 | 8069002 |