Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SGIP1 SIRNA Assays ›
          Gene Symbol:
          SGIP1
          Gene Name:
          SH3 domain GRB2 like endophilin interacting protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 66533272 - 66751139 on Build GRCh38
          UniGene ID:
          Hs.132121
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS130805
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SGIP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001308203.1 NP_001295132.1 20, 21 1936
          NM_032291.3 NP_115667.2 23, 24 2713
          XM_005271264.3 XP_005271321.1 23, 24 2547
          XM_005271268.3 XP_005271325.1 21, 22 1962
          XM_005271270.4 XP_005271327.1 20, 21 2269
          XM_006710961.2 XP_006711024.1 23, 24 2553
          XM_006710966.2 XP_006711029.1 22, 23 2487
          XM_006710967.2 XP_006711030.1 22, 23 2475
          XM_006710969.2 XP_006711032.1 22, 23 2463
          XM_006710971.2 XP_006711034.1 22, 23 2052
          XM_006710972.2 XP_006711035.1 22, 23 2046
          XM_006710973.2 XP_006711036.1 20, 21 2028
          XM_006710974.2 XP_006711037.1 19, 20 1944
          XM_011542291.1 XP_011540593.1 22, 23 2209
          XM_011542292.1 XP_011540594.1 21, 22 2149
          XM_011542293.1 XP_011540595.1 18, 19 1878
          XM_017002505.1 XP_016857994.1 24, 25 2601
          XM_017002506.1 XP_016857995.1 25, 26 2724
          XM_017002507.1 XP_016857996.1 24, 25 2776
          XM_017002508.1 XP_016857997.1 25, 26 2722
          XM_017002509.1 XP_016857998.1 22, 23 2577
          XM_017002510.1 XP_016857999.1 23, 24 2700
          XM_017002511.1 XP_016858000.1 24, 25 2658
          XM_017002512.1 XP_016858001.1 24, 25 2531
          XM_017002513.1 XP_016858002.1 23, 24 2710
          XM_017002514.1 XP_016858003.1 23, 24 2529
          XM_017002515.1 XP_016858004.1 21, 22 2505
          XM_017002516.1 XP_016858005.1 22, 23 2628
          XM_017002517.1 XP_016858006.1 23, 24 2597
          XM_017002518.1 XP_016858007.1 23, 24 2585
          XM_017002519.1 XP_016858008.1 23, 24 2610
          XM_017002520.1 XP_016858009.1 21, 22 2451
          XM_017002521.1 XP_016858010.1 21, 22 2403
          XM_017002522.1 XP_016858011.1 22, 23 2525
          XM_017002523.1 XP_016858012.1 22, 23 2549
          XM_017002524.1 XP_016858013.1 22, 23 2192
          XM_017002525.1 XP_016858014.1 21, 22 2131
          XM_017002526.1 XP_016858015.1 20, 21 2010
          XM_017002527.1 XP_016858016.1 21, 22 2034
          XM_017002528.1 XP_016858017.1 21, 22 2120
          XM_017002529.1 XP_016858018.1 21, 22 1986
          XM_017002530.1 XP_016858019.1 20, 21 1962
          XM_017002531.1 XP_016858020.1 19, 20 1962
          XM_017002532.1 XP_016858021.1 20, 21 1956
          XM_017002533.1 XP_016858022.1 20, 21 2064
          XM_017002534.1 XP_016858023.1 19, 20 1938
          XM_017002535.1 XP_016858024.1 20, 21 2052
          XM_017002536.1 XP_016858025.1 20, 21 1902
          XM_017002537.1 XP_016858026.1 19, 20 1991
          GenBank mRNA AB210039.1 - 23, 24 2495
          AK090697.1 - 14, 15 1493
          AK125044.1 - 6, 7 1480
          AK298970.1 - 20, 21 1899
          AK299941.1 - 21, 22 1973
          AL136561.1 - 23, 24 2387
          BC040516.1 - 25, 26 2541 5275011
          BX640813.1 - 20, 21 1913

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          positive regulation of receptor-mediated endocytosis
          membrane tubulation
          protein binding
          phospholipid binding
          microtubule binding
          SH3 domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-s6kwc:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0