Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGIP1 SIRNA Assays ›
Gene Symbol:
SGIP1
Gene Name:
SH3 domain GRB2 like endophilin interacting protein 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 66533272 - 66751139 on Build GRCh38
UniGene ID:
Hs.132121
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS130806
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308203.1
NP_001295132.1
21
1976
NM_032291.3
NP_115667.2
24
2753
XM_005271264.3
XP_005271321.1
24
2587
XM_005271268.3
XP_005271325.1
22
2002
XM_005271270.4
XP_005271327.1
21
2309
XM_006710961.2
XP_006711024.1
24
2593
XM_006710966.2
XP_006711029.1
23
2527
XM_006710967.2
XP_006711030.1
23
2515
XM_006710969.2
XP_006711032.1
23
2503
XM_006710971.2
XP_006711034.1
23
2092
XM_006710972.2
XP_006711035.1
23
2086
XM_006710973.2
XP_006711036.1
21
2068
XM_006710974.2
XP_006711037.1
20
1984
XM_011542291.1
XP_011540593.1
23
2249
XM_011542292.1
XP_011540594.1
22
2189
XM_011542293.1
XP_011540595.1
19
1918
XM_017002505.1
XP_016857994.1
25
2641
XM_017002506.1
XP_016857995.1
26
2764
XM_017002507.1
XP_016857996.1
25
2816
XM_017002508.1
XP_016857997.1
26
2762
XM_017002509.1
XP_016857998.1
23
2617
XM_017002510.1
XP_016857999.1
24
2740
XM_017002511.1
XP_016858000.1
25
2698
XM_017002512.1
XP_016858001.1
25
2571
XM_017002513.1
XP_016858002.1
24
2750
XM_017002514.1
XP_016858003.1
24
2569
XM_017002515.1
XP_016858004.1
22
2545
XM_017002516.1
XP_016858005.1
23
2668
XM_017002517.1
XP_016858006.1
24
2637
XM_017002518.1
XP_016858007.1
24
2625
XM_017002519.1
XP_016858008.1
24
2650
XM_017002520.1
XP_016858009.1
22
2491
XM_017002521.1
XP_016858010.1
22
2443
XM_017002522.1
XP_016858011.1
23
2565
XM_017002523.1
XP_016858012.1
23
2589
XM_017002524.1
XP_016858013.1
23
2232
XM_017002525.1
XP_016858014.1
22
2171
XM_017002526.1
XP_016858015.1
21
2050
XM_017002527.1
XP_016858016.1
22
2074
XM_017002528.1
XP_016858017.1
22
2160
XM_017002529.1
XP_016858018.1
22
2026
XM_017002530.1
XP_016858019.1
21
2002
XM_017002531.1
XP_016858020.1
20
2002
XM_017002532.1
XP_016858021.1
21
1996
XM_017002533.1
XP_016858022.1
21
2104
XM_017002534.1
XP_016858023.1
20
1978
XM_017002535.1
XP_016858024.1
21
2092
XM_017002536.1
XP_016858025.1
21
1942
XM_017002537.1
XP_016858026.1
20
2031
GenBank mRNA
AB210039.1
-
24
2535
AK090697.1
-
15
1533
AK125044.1
-
7
1520
AK298970.1
-
21
1939
AK299941.1
-
22
2013
AL136561.1
-
24
2427
BC040516.1
-
26
2581
5275011
BX640813.1
-
21
1953
CR749541.1
-
21
2164
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
positive regulation of receptor-mediated endocytosis
membrane tubulation
protein binding
phospholipid binding
microtubule binding
SH3 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0