Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCDC115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321118.1 | NP_001308047.1 | 5 | 901 | ||
NM_001321119.1 | NP_001308048.1 | 5 | 423 | |||
NM_032357.3 | NP_115733.2 | 5 | 722 | |||
NR_104471.1 | 6 | 1104 | ||||
NR_135548.1 | 6 | 925 | ||||
GenBank mRNA | AK054693.1 | - | 5 | 688 | ||
AK092196.1 | - | 1 | 1425 | |||
AK124405.1 | - | 2 | 2610 | |||
AK295922.1 | - | 6 | 761 | |||
BC006429.1 | - | 5 | 484 | 3350642 | ||
BG179982.1 | - | 423 |