Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027128.1 | 2 | 437 | |||
NR_027129.1 | 3 | 581 | ||||
GenBank mRNA | AF380178.1 | - | 3 | 513 | ||
AY040088.1 | - | 3 | 513 | |||
BC128244.1 | - | 2 | 295 | |||
BC128245.1 | - | 2 | 295 | |||
BI829356.1 | - | 2 | 401 | |||
BI907880.1 | - | 2 | 437 |