Search
Search

ZBTB37| Gene Symbol: ZBTB37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122770.1 | NP_001116242.1 | 3 | 373 | ||
| NM_032522.3 | NP_115911.1 | 3 | 370 | |||
| XM_005245546.2 | XP_005245603.1 | 3 | 428 | |||
| XM_006711578.3 | XP_006711641.1 | 2 | 437 | |||
| XM_011510062.2 | XP_011508364.1 | 3 | 428 | |||
| XM_017002556.1 | XP_016858045.1 | 2 | 438 | |||
| XM_017002557.1 | XP_016858046.1 | 3 | 348 | |||
| XM_017002558.1 | XP_016858047.1 | 3 | 428 | |||
| GenBank mRNA | AF318371.1 | - | 2 | 289 | ||
| AK057310.1 | - | 3 | 496 | |||
| AK302696.1 | - | 3 | 345 | |||
| BC003116.1 | - | 2 | 331 | 3503643 | ||
| BC006315.2 | - | 2 | 331 | 4124255 | ||
| DA790498.1 | - | 3 | 348 | |||
| DA796002.1 | - | 3 | 370 | |||