Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256267.1 | NP_001243196.1 | 9, 10 | 1677 | ||
NM_001256268.1 | NP_001243197.1 | 12, 13 | 2086 | |||
NM_032578.3 | NP_115967.2 | 8, 9 | 1782 | |||
NR_045662.3 | 8, 9 | 917 | ||||
XM_017016833.1 | XP_016872322.1 | 9, 10 | 2118 | |||
XM_017016834.1 | XP_016872323.1 | 8, 9 | 1576 | |||
GenBank mRNA | AF328296.1 | - | 8, 9 | 1977 | ||
AL832002.2 | - | 8, 9 | 1705 | |||
AL832379.1 | - | 8, 9 | 879 | |||
AL834247.2 | - | 8, 9 | 1704 | |||
BC142609.1 | - | 8, 9 | 1654 | |||
BX647637.1 | - | 8, 9 | 1714 | |||
BX647640.1 | - | 9, 10 | 1670 | |||
BX648564.1 | - | 10, 11 | 2110 |