Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257291.1 | NP_001244220.1 | 12 | 1674 | ||
NM_032591.2 | NP_115980.1 | 12 | 1671 | |||
XM_005272681.4 | XP_005272738.1 | 12 | 1664 | |||
XM_005272682.4 | XP_005272739.1 | 12 | 1664 | |||
XM_006724564.3 | XP_006724627.1 | 12 | 1664 | |||
XM_011543990.2 | XP_011542292.1 | 12 | 1664 | |||
XM_017029905.1 | XP_016885394.1 | 12 | 1664 | |||
XM_017029906.1 | XP_016885395.1 | 12 | 1664 | |||
GenBank mRNA | AF298591.1 | - | 12 | 1536 | ||
AK056599.1 | - | 10 | 999 |