Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136493.2 | NP_001129965.1 | 13 | 1611 | ||
NM_001287808.1 | NP_001274737.1 | 11 | 1312 | |||
NM_001287809.1 | NP_001274738.1 | 13 | 1455 | |||
NM_032793.4 | NP_116182.2 | 13 | 1572 | |||
NR_109896.1 | 11 | 1375 | ||||
XM_005271285.1 | XP_005271342.1 | 13 | 1536 | |||
XM_011542312.2 | XP_011540614.1 | 12 | 1455 | |||
GenBank mRNA | AB073383.1 | - | 13 | 1536 | ||
AF289609.1 | - | 13 | 1528 | |||
AF370364.1 | - | 8 | 1832 | |||
AK074186.1 | - | 13 | 1477 | |||
AK075183.1 | - | 13 | 1539 | |||
AK093223.1 | - | 13 | 1573 | |||
AK291540.1 | - | 13 | 1534 | |||
AK297991.1 | - | 13 | 1419 | |||
AK301533.1 | - | 11 | 1279 | |||
AL834184.1 | - | 11 | 1329 | |||
AY358636.1 | - | 13 | 1554 | |||
BC006353.2 | - | 9 | 907 | |||
BC011587.1 | - | 13 | 1546 | 4646430 | ||
BC092414.1 | - | 13 | 1514 | 30387073 |