Search
Search

RPUSD4| Gene Symbol: RPUSD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144827.1 | NP_001138299.1 | 7 | 994 | ||
| NM_032795.2 | NP_116184.2 | 7 | 1087 | |||
| XM_011543039.2 | XP_011541341.1 | 7 | 1084 | |||
| GenBank mRNA | AK027400.1 | - | 7 | 1041 | ||
| AK225320.1 | - | 7 | 1047 | |||
| AK300723.1 | - | 5 | 561 | |||
| AK308059.1 | - | 7 | 947 | |||
| BC014131.2 | - | 7 | 1048 | 4621434 | ||
| BE260782.1 | - | 1 | 85 | |||