Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PLXDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282736.1 | NP_001269665.1 | 4 | 1084 | ||
NM_032812.8 | NP_116201.7 | 5 | 1231 | |||
XM_011519750.2 | XP_011518052.1 | 5 | 1459 | |||
GenBank mRNA | AF378757.1 | - | 5 | 603 | ||
AK304591.1 | - | 6 | 1115 | |||
AY358486.1 | - | 5 | 1231 | |||
BC012885.1 | - | 4 | 690 | 3865073 |