Search
Search

CEP19| Gene Symbol: CEP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032898.4 | NP_116287.2 | 2 | 535 | ||
| XM_005269370.4 | XP_005269427.1 | 2 | 510 | |||
| XM_011513246.2 | XP_011511548.1 | 2 | 271 | |||
| GenBank mRNA | AK058155.1 | - | 2 | 438 | ||
| AK314070.1 | - | 2 | 504 | |||
| AY099509.1 | - | 2 | 465 | |||
| BC007827.2 | - | 2 | 464 | 3533603 | ||