Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173462.3 | NP_775733.3 | 26 | 3779 | ||
XM_011537290.2 | XP_011535592.1 | 27 | 3860 | |||
XM_011537291.2 | XP_011535593.1 | 28 | 3907 | |||
XM_011537292.2 | XP_011535594.1 | 27 | 3783 | |||
XM_011537293.2 | XP_011535595.1 | 26 | 3826 | |||
XM_011537294.2 | XP_011535596.1 | 26 | 3784 | |||
XM_011537295.2 | XP_011535597.1 | 24 | 3542 | |||
XM_017021749.1 | XP_016877238.1 | 24 | 3589 | |||
XM_017021750.1 | XP_016877239.1 | 26 | 3782 | |||
GenBank mRNA | AK125658.1 | - | 6 | 959 | ||
AK127354.1 | - | 12 | 1928 | |||
AK131073.1 | - | 19 | 3072 | |||
AK293773.1 | - | 26 | 3921 | |||
AK294560.1 | - | 26 | 3864 | |||
AL110280.1 | - | 8 | 2631 | |||
AY358330.1 | - | 10 | 1684 | |||
BC136623.1 | - | 26 | 3403 | 9020613 | ||
BC144315.1 | - | 23 | 3328 | |||
BX537757.1 | - | 12 | 1905 |