Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138342.3 | NP_612351.2 | 15, 16 | 1700 | ||
XM_006718937.1 | XP_006719000.1 | 15, 16 | 1640 | |||
GenBank mRNA | AF173889.1 | - | 14, 15 | 1327 | ||
AK074750.1 | - | 6, 7 | 536 | |||
AK074855.1 | - | 15, 16 | 1570 | |||
AK074947.1 | - | 15, 16 | 1524 | |||
AY358497.1 | - | 15, 16 | 1580 | |||
BC008326.1 | - | 11, 12 | 987 | |||
BC040641.1 | - | 15, 16 | 1662 | 3849776 |