Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WDR20 SIRNA Assays ›
Gene Symbol:
WDR20
Gene Name:
WD repeat domain 20
Gene Aliases:
DMR
Chromosome Location:
Chr.14: 102139518 - 102224686 on Build GRCh38
UniGene ID:
Hs.36859
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS132257
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WDR20
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242414.1
NP_001229343.1
3
1283
NM_001242416.1
NP_001229345.1
2
1270
NM_001242417.1
NP_001229346.1
4
1546
NM_001242418.1
NP_001229347.1
3
1489
NM_144574.3
NP_653175.2
3
1453
NM_181291.2
NP_851808.1
3
1453
NM_181308.2
NP_851825.1
2
1270
NR_038354.1
4
1246
XM_006720308.1
XP_006720371.1
2
1262
XM_006720309.1
XP_006720372.1
3
1236
XM_006720310.1
XP_006720373.1
3
1197
XM_006720311.1
XP_006720374.1
2
1026
XM_006720313.2
XP_006720376.1
4
1205
XM_006720318.2
XP_006720381.1
3
1144
XM_011537335.2
XP_011535637.1
4
1565
XM_011537336.2
XP_011535638.1
4
1565
XM_011537337.2
XP_011535639.1
4
1565
XM_011537338.1
XP_011535640.1
4
1512
XM_011537339.2
XP_011535641.1
4
1565
XM_011537340.2
XP_011535642.1
4
1565
XM_011537341.2
XP_011535643.1
3
1471
XM_011537343.1
XP_011535645.1
5
1540
XM_011537344.2
XP_011535646.1
4
1458
XM_011537345.1
XP_011535647.1
3
1236
XM_011537346.1
XP_011535648.1
3
1262
XM_011537347.1
XP_011535649.1
4
1260
XM_011537353.2
XP_011535655.1
4
1316
XM_011537358.2
XP_011535660.1
3
1221
XM_017021766.1
XP_016877255.1
4
1512
XM_017021767.1
XP_016877256.1
3
1471
XM_017021768.1
XP_016877257.1
3
1486
XM_017021770.1
XP_016877259.1
3
1486
XM_017021771.1
XP_016877260.1
4
1521
XM_017021772.1
XP_016877261.1
4
1274
XM_017021773.1
XP_016877262.1
5
1367
XM_017021774.1
XP_016877263.1
4
1316
XM_017021775.1
XP_016877264.1
4
1322
XM_017021776.1
XP_016877265.1
3
1236
XM_017021777.1
XP_016877266.1
3
1204
XM_017021778.1
XP_016877267.1
4
1316
XM_017021779.1
XP_016877268.1
2
1026
XM_017021780.1
XP_016877269.1
4
1437
XM_017021781.1
XP_016877270.1
3
1144
XM_017021782.1
XP_016877271.1
3
957
XM_017021783.1
XP_016877272.1
4
1253
XM_017021785.1
XP_016877274.1
4
1512
GenBank mRNA
AB209148.1
-
1
5542
AF327354.1
-
3
1378
AK056330.1
-
3
1195
AK090580.1
-
4
1205
AK090978.1
-
3
1224
AK225852.1
-
2
1229
AK297727.1
-
4
1484
AL133558.1
-
1
690
AL832242.1
-
3
1222
AL832500.1
-
3
1428
BC028387.2
-
3
1412
4823662
BX248274.1
-
2
1172
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-22h96:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0