Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174889.4 | NP_777549.1 | 1 | 147 | ||
GenBank mRNA | AB183433.1 | - | 1 | 20 | ||
AF087990.1 | - | 1 | 116 | |||
AK026051.1 | - | 1 | 147 | |||
AK291296.1 | - | 1 | 110 | |||
BC001753.1 | - | 1 | 69 | 3354845 | ||
BC022869.1 | - | 1 | 48 | |||
BC033965.1 | - | 1 | 63 | 5288040 | ||
BC070357.1 | - | 1 | 69 | |||
KU178834.1 | - | 1 | 20 | |||
KU178835.1 | - | 1 | 20 | |||
KU178836.1 | - | 1 | 20 |