Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FANK1 SIRNA Assays ›
Gene Symbol:
FANK1
Gene Name:
fibronectin type III and ankyrin repeat domains 1
Gene Aliases:
HSD13
Chromosome Location:
Chr.10: 125896539 - 126009599 on Build GRCh38
UniGene ID:
Hs.352591
Species:
Human
siRNA ID
HSS132371
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_145235.3
NP_660278.3
6, 7
648
XM_005270280.3
XP_005270337.1
5, 6
464
XM_006718058.3
XP_006718121.1
8, 9
647
XM_006718064.3
XP_006718127.1
7, 8
641
XM_011540348.2
XP_011538650.1
8, 9
762
XM_011540349.2
XP_011538651.1
8, 9
766
XM_011540350.2
XP_011538652.1
7, 8
725
XM_011540351.2
XP_011538653.1
7, 8
701
XM_011540356.2
XP_011538658.1
6, 7
613
XM_011540357.2
XP_011538659.1
8, 9
709
XM_011540358.2
XP_011538660.1
6, 7
628
XM_011540360.2
XP_011538662.1
6, 7
542
XM_011540361.2
XP_011538663.1
6, 7
477
XM_011540362.2
XP_011538664.1
4, 5
363
XM_017016916.1
XP_016872405.1
8, 9
708
XM_017016917.1
XP_016872406.1
8, 9
843
XM_017016918.1
XP_016872407.1
7, 8
650
XM_017016919.1
XP_016872408.1
7, 8
802
XM_017016921.1
XP_016872410.1
7, 8
765
XM_017016922.1
XP_016872411.1
7, 8
665
XM_017016923.1
XP_016872412.1
6, 7
647
XM_017016924.1
XP_016872413.1
9, 10
820
XM_017016925.1
XP_016872414.1
5, 6
7370
XM_017016926.1
XP_016872415.1
3, 4
777
GenBank mRNA
AY251163.1
-
5, 6
548
AY358154.1
-
7, 8
704
BC024189.2
-
6, 7
648
4826296
BQ225468.1
-
6, 7
592
CR627249.1
-
5, 6
7362
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-c74kb:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0