Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPH7 SIRNA Assays ›
Gene Symbol:
DPH7
Gene Name:
diphthamide biosynthesis 7
Gene Aliases:
C9orf112, RRT2, WDR85
Chromosome Location:
Chr.9: 137554899 - 137578935 on Build GRCh38
UniGene ID:
Hs.292570
Species:
Human
siRNA ID
HSS132408
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPH7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_138778.3
NP_620133.1
5
734
XM_005266123.2
XP_005266180.1
6
1192
XM_005266126.3
XP_005266183.1
4
965
XM_005266128.2
XP_005266185.1
6
818
XM_005266129.2
XP_005266186.1
6
821
XM_005266130.2
XP_005266187.1
4
589
XM_011519188.1
XP_011517490.1
5
723
XM_011519189.1
XP_011517491.1
5
835
XM_011519190.2
XP_011517492.1
5
723
XM_011519191.2
XP_011517493.1
5
723
XM_011519192.2
XP_011517494.1
5
723
XM_011519193.2
XP_011517495.1
5
723
XM_011519194.1
XP_011517496.1
5
723
XM_011519199.2
XP_011517501.1
5
680
XM_011519200.1
XP_011517502.1
5
683
XM_017015289.1
XP_016870778.1
5
723
XM_017015290.1
XP_016870779.1
5
723
XM_017015291.1
XP_016870780.1
6
1027
XM_017015292.1
XP_016870781.1
5
1097
XM_017015293.1
XP_016870782.1
5
1054
XM_017015294.1
XP_016870783.1
7
1007
XM_017015295.1
XP_016870784.1
5
1094
XM_017015296.1
XP_016870785.1
5
933
XM_017015297.1
XP_016870786.1
7
1004
XM_017015298.1
XP_016870787.1
6
1287
XM_017015299.1
XP_016870788.1
6
544
XM_017015300.1
XP_016870789.1
6
1284
XM_017015301.1
XP_016870790.1
4
957
XM_017015302.1
XP_016870791.1
4
2011
XM_017015303.1
XP_016870792.1
5
1192
XM_017015304.1
XP_016870793.1
4
657
XM_017015305.1
XP_016870794.1
5
2054
XM_017015306.1
XP_016870795.1
6
1192
XM_017015307.1
XP_016870796.1
6
1189
XM_017015308.1
XP_016870797.1
4
1916
XM_017015309.1
XP_016870798.1
6
916
GenBank mRNA
AK075115.1
-
5
735
AK308998.1
-
4
589
BC003123.1
-
5
673
3502107
BC017335.2
-
5
693
4642600
Back To Top
More Information
Panther Classification:
Molecular Function -
methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-diphthamide biosynthetic process from peptidyl-histidine
diphthine methylesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-2px8h:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0