Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320304.1 | NP_001307233.1 | 6 | 1206 | ||
NM_001320305.1 | NP_001307234.1 | 6 | 1158 | |||
NM_001320306.1 | NP_001307235.1 | 6 | 1225 | |||
NM_001320307.1 | NP_001307236.1 | 6 | 1252 | |||
NM_001320309.1 | NP_001307238.1 | 5 | 1362 | |||
NM_001320310.1 | NP_001307239.1 | 6 | 1239 | |||
NM_001320311.1 | NP_001307240.1 | 6 | 1230 | |||
NM_033416.2 | NP_219484.1 | 6 | 1212 | |||
GenBank mRNA | AF054996.1 | - | 6 | 1027 | ||
AK289878.1 | - | 6 | 540 | |||
AK292345.1 | - | 6 | 540 | |||
AK312808.1 | - | 6 | 540 | |||
AY364245.1 | - | 5 | 504 | |||
BC010042.1 | - | 6 | 521 | 3629513 | ||
DA893334.1 | - | 6 | 508 | |||
DB200539.1 | - | 6 | 537 |