Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFAND4 SIRNA Assays ›
Gene Symbol:
ZFAND4
Gene Name:
zinc finger AN1-type containing 4
Gene Aliases:
ANUBL1
Chromosome Location:
Chr.10: 45614567 - 45672962 on Build GRCh38
UniGene ID:
Hs.659717
Species:
Human
siRNA ID
HSS132542
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFAND4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128324.2
NP_001121796.1
7
1827
NM_001282905.1
NP_001269834.1
8
2277
NM_001282906.1
NP_001269835.1
8
1929
NM_174890.3
NP_777550.2
7
2175
XM_005271842.2
XP_005271899.1
6
1700
XM_005271843.2
XP_005271900.1
6
1867
XM_005271844.3
XP_005271901.1
4
1333
XM_011540366.2
XP_011538668.1
5
1771
XM_011540368.1
XP_011538670.1
4
1216
XM_017016931.1
XP_016872420.1
5
1791
XM_017016932.1
XP_016872421.1
7
1937
XM_017016933.1
XP_016872422.1
7
1804
XM_017016934.1
XP_016872423.1
7
1857
XM_017016935.1
XP_016872424.1
6
1607
XM_017016936.1
XP_016872425.1
5
1947
XM_017016937.1
XP_016872426.1
5
1795
XM_017016938.1
XP_016872427.1
5
2089
XM_017016939.1
XP_016872428.1
5
1660
XM_017016940.1
XP_016872429.1
8
2353
XM_017016941.1
XP_016872430.1
8
1896
XM_017016942.1
XP_016872431.1
6
1885
XM_017016943.1
XP_016872432.1
6
2161
XM_017016944.1
XP_016872433.1
6
1718
XM_017016945.1
XP_016872434.1
4
1305
XM_017016946.1
XP_016872435.1
8
1971
GenBank mRNA
AF311324.1
-
7
1791
AK097504.1
-
4
1295
AK292420.1
-
8
1878
AK292469.1
-
7
1776
AK314395.1
-
7
1787
BC045587.1
-
7
1827
5269483
BC048968.1
-
8
1819
5272133
Back To Top
More Information
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-bsnw9:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0