Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CADPS2 SIRNA Assays ›
Gene Symbol:
CADPS2
Gene Name:
calcium dependent secretion activator 2
Gene Aliases:
CAPS2
Chromosome Location:
Chr.7: 122318424 - 122886759 on Build GRCh38
UniGene ID:
Hs.649459
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS132567
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CADPS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001009571.3
NP_001009571.2
10, 11
2083
NM_001167940.1
NP_001161412.1
10, 11
2083
NM_017954.10
NP_060424.9
10, 11
2083
XM_005250695.4
XP_005250752.1
10, 11
1812
XM_005250696.4
XP_005250753.1
10, 11
1812
XM_005250697.4
XP_005250754.1
10, 11
1812
XM_005250698.4
XP_005250755.1
10, 11
1784
XM_005250699.4
XP_005250756.1
10, 11
1812
XM_005250700.4
XP_005250757.1
10, 11
1812
XM_005250701.4
XP_005250758.1
10, 11
1812
XM_005250702.4
XP_005250759.1
10, 11
1812
XM_005250703.4
XP_005250760.1
10, 11
1812
XM_005250704.4
XP_005250761.1
10, 11
1812
XM_005250705.4
XP_005250762.1
10, 11
1812
XM_005250706.4
XP_005250763.1
10, 11
1812
XM_005250707.4
XP_005250764.1
10, 11
1812
XM_005250708.3
XP_005250765.1
10, 11
1812
XM_011516695.2
XP_011514997.1
10, 11
1784
XM_011516696.2
XP_011514998.1
10, 11
1812
XM_011516697.2
XP_011514999.1
10, 11
1812
XM_017012794.1
XP_016868283.1
10, 11
1812
XM_017012795.1
XP_016868284.1
10, 11
1812
XM_017012796.1
XP_016868285.1
11, 12
1427
XM_017012797.1
XP_016868286.1
11, 12
1493
XM_017012798.1
XP_016868287.1
10, 11
1963
XM_017012799.1
XP_016868288.1
11, 12
1864
XM_017012800.1
XP_016868289.1
7, 8
820
GenBank mRNA
AB046811.1
-
8, 9
952
AF401638.1
-
10, 11
1752
AK025672.1
-
6, 7
606
AK126518.1
-
11, 12
1685
AL833058.1
-
7, 8
878
AY264289.1
-
10, 11
1687
BC136601.1
-
10, 11
1782
9020591
BC144278.1
-
10, 11
1782
9052798
BC144279.1
-
10, 11
1782
9052799
KJ534798.1
-
10, 11
1661
KJ534800.1
-
10, 11
1661
KJ534801.1
-
10, 11
1661
KJ534802.1
-
10, 11
1661
KJ534803.1
-
10, 11
1661
KJ534804.1
-
10, 11
1661
KJ534805.1
-
10, 11
1661
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
cellular response to starvation
protein transport
synaptic vesicle priming
positive regulation of exocytosis
dense core granule exocytosis
lipid binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0