Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256605.1 | NP_001243534.1 | 4 | 711 | ||
NM_001256609.1 | NP_001243538.1 | 5 | 1198 | |||
NM_144580.2 | NP_653181.1 | 5 | 969 | |||
XM_011509117.2 | XP_011507419.1 | 4 | 628 | |||
GenBank mRNA | AK075349.1 | - | 5 | 938 | ||
AK296157.1 | - | 5 | 1166 | |||
AL528533.3 | - | 4 | 683 | |||
AY358450.1 | - | 5 | 942 | |||
BC011575.1 | - | 5 | 952 | |||
BC018757.1 | - | 5 | 938 | 4850051 | ||
BC036340.1 | - | 5 | 885 | |||
BM833126.1 | - | 3 | 371 |