Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OSBPL6 SIRNA Assays ›
Gene Symbol:
OSBPL6
Gene Name:
oxysterol binding protein like 6
Gene Aliases:
ORP6
Chromosome Location:
Chr.2: 178194479 - 178399433 on Build GRCh38
UniGene ID:
Hs.318775
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS132904
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OSBPL6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001201480.1
NP_001188409.1
8, 9
1205
NM_001201481.1
NP_001188410.1
8, 9
1205
NM_001201482.1
NP_001188411.1
8, 9
1205
NM_032523.3
NP_115912.1
8, 9
1205
NM_145739.2
NP_665682.1
6, 7
718
XM_011510539.1
XP_011508841.1
7, 8
1108
XM_017003265.1
XP_016858754.1
9, 10
1277
XM_017003266.1
XP_016858755.1
9, 10
1277
XM_017003267.1
XP_016858756.1
7, 8
972
XM_017003268.1
XP_016858757.1
8, 9
1201
XM_017003269.1
XP_016858758.1
7, 8
972
XM_017003270.1
XP_016858759.1
9, 10
1359
XM_017003271.1
XP_016858760.1
7, 8
972
XM_017003272.1
XP_016858761.1
7, 8
1108
XM_017003273.1
XP_016858762.1
7, 8
1108
XM_017003274.1
XP_016858763.1
8, 9
1201
XM_017003275.1
XP_016858764.1
9, 10
1295
XM_017003276.1
XP_016858765.1
7, 8
1108
XM_017003277.1
XP_016858766.1
7, 8
1108
GenBank mRNA
AB208898.1
-
8, 9
1205
AF323728.1
-
8, 9
1039
AF392448.1
-
8, 9
1039
AF462443.1
-
7, 8
1015
AK027600.1
-
1, 2
80
AK093902.1
-
6, 7
720
AK300389.1
-
8, 9
1163
BC021248.2
-
17
BC052259.1
-
8, 9
1186
6428611
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
lipid transport
protein binding
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-6tb5w:80/100.66.72.8:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0