Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052960.2 | NP_443192.1 | 3 | 338 | ||
GenBank mRNA | AF399927.1 | - | 3 | 319 | ||
AK312027.1 | - | 3 | 306 | |||
AY145438.1 | - | 3 | 305 | |||
BC033883.1 | - | 4 | 551 | |||
BC063013.1 | - | 3 | 338 | 5224517 |