Search
Search

CPXM2| Gene Symbol: CPXM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198148.2 | NP_937791.2 | 4, 5 | 986 | ||
| XM_005269528.3 | XP_005269585.1 | 4, 5 | 836 | |||
| XM_011539283.2 | XP_011537585.1 | 4, 5 | 836 | |||
| XM_011539285.2 | XP_011537587.1 | 4, 5 | 618 | |||
| XM_011539286.1 | XP_011537588.1 | 3, 4 | 560 | |||
| XM_017015673.1 | XP_016871162.1 | 5, 6 | 587 | |||
| GenBank mRNA | AK296365.1 | - | 4, 5 | 393 | ||
| AK304023.1 | - | 4, 5 | 760 | |||
| AK304814.1 | - | 4, 5 | 797 | |||
| AL833732.1 | - | 2, 3 | 164 | |||
| AY358565.1 | - | 4, 5 | 986 | |||
| BC027623.1 | - | 5, 6 | 532 | |||
| BC036789.1 | - | 4, 5 | 816 | 5752707 | ||
| BC137492.1 | - | 4, 5 | 981 | 9021492 | ||
| BC146862.1 | - | 4, 5 | 981 | 9056790 | ||