Search
Search

ARL14EP| Gene Symbol: ARL14EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152316.2 | NP_689529.1 | 4 | 820 | ||
| XM_005252792.3 | XP_005252849.1 | 4 | 1271 | |||
| GenBank mRNA | AK096287.1 | - | 4 | 817 | ||
| BC047775.1 | - | 4 | 808 | 6062049 | ||
| BX647617.1 | - | 4 | 813 | |||