Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM52B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079815.1 | NP_001073283.1 | 5 | 556 | ||
NM_153022.3 | NP_694567.1 | 3 | 782 | |||
XM_006719029.2 | XP_006719092.1 | 4 | 826 | |||
GenBank mRNA | AK055728.1 | - | 3 | 708 | ||
AY358845.1 | - | 3 | 739 | |||
BC098571.1 | - | 4 | 266 | 30921470 | ||
BC131523.1 | - | 4 | 270 | 40032531 |