Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152324.2 | NP_689537.1 | 3, 4 | 323 | ||
XM_017020387.1 | XP_016875876.1 | 3, 4 | 1631 | |||
XM_017020388.1 | XP_016875877.1 | 4, 5 | 485 | |||
GenBank mRNA | BC029889.1 | - | 3, 4 | 299 | 5170095 | |
HY035354.1 | - | 3, 4 | 324 |