Search
Search

ADSSL1| Gene Symbol: ADSSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320424.1 | NP_001307353.1 | 9 | 973 | ||
| NM_152328.4 | NP_689541.1 | 9 | 886 | |||
| NM_199165.2 | NP_954634.1 | 9 | 990 | |||
| XM_006720026.3 | XP_006720089.1 | 10 | 885 | |||
| XM_011536412.1 | XP_011534714.1 | 10 | 959 | |||
| XM_011536414.2 | XP_011534716.1 | 10 | 642 | |||
| GenBank mRNA | AK095921.1 | - | 9 | 882 | ||
| AK096124.1 | - | 7 | 1733 | |||
| AK096720.1 | - | 9 | 924 | |||
| AY037159.1 | - | 9 | 871 | |||
| BC032039.1 | - | 9 | 947 | |||
| BC047904.1 | - | 9 | 886 | 5755025 | ||
| BX248286.1 | - | 9 | 843 | |||