Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152335.2 | NP_689548.2 | 7, 8 | 797 | ||
XM_005254160.2 | XP_005254217.1 | 7, 8 | 720 | |||
XM_017021915.1 | XP_016877404.1 | 9, 10 | 1013 | |||
XM_017021916.1 | XP_016877405.1 | 9, 10 | 959 | |||
XM_017021917.1 | XP_016877406.1 | 9, 10 | 945 | |||
GenBank mRNA | AK095509.1 | - | 7, 8 | 689 | ||
AK308676.1 | - | 6, 7 | 615 | |||
BC104953.1 | - | 1, 2 | 124 | 8143956 | ||
BC104955.1 | - | 1, 2 | 124 | 8143958 | ||
DA483185.1 | - | 7, 8 | 689 | |||
DR007145.1 | - | 3, 4 | 267 |