Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145205.5 | NP_660206.2 | 2 | 1871 | ||
NR_033264.1 | 2 | 156 | ||||
GenBank mRNA | AK097707.1 | - | 2 | 1872 | ||
AK130859.1 | - | 1 | 290 | |||
AK310465.1 | - | 2 | 156 | |||
AK311985.1 | - | 1 | 257 | |||
BC021180.2 | - | 1 | 241 | 5266418 | ||
BC070148.1 | - | 2 | 156 | 6617009 | ||
BI827031.1 | - | 1 | 230 | |||
DQ207367.1 | - | 1 | 241 |