Search
Search

RBM45| Gene Symbol: RBM45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152945.3 | NP_694453.2 | 3 | 617 | ||
| XM_005246287.4 | XP_005246344.1 | 3 | 627 | |||
| XM_017003320.1 | XP_016858809.1 | 3 | 1060 | |||
| XM_017003321.1 | XP_016858810.1 | 3 | 1061 | |||
| XM_017003322.1 | XP_016858811.1 | 3 | 626 | |||
| GenBank mRNA | AB036991.1 | - | 3 | 539 | ||
| AF526533.1 | - | 3 | 617 | |||
| AK126575.1 | - | 3 | 475 | |||
| BC066549.1 | - | 3 | 586 | 5748065 | ||