Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172070.3 | NP_742067.3 | 32, 33 | 4663 | ||
XM_005246299.4 | XP_005246356.1 | 33, 34 | 4753 | |||
XM_005246300.4 | XP_005246357.1 | 33, 34 | 4696 | |||
XM_006712268.3 | XP_006712331.1 | 33, 34 | 4750 | |||
XM_006712269.3 | XP_006712332.1 | 32, 33 | 4675 | |||
XM_006712270.3 | XP_006712333.2 | 32, 33 | 4609 | |||
XM_006712271.3 | XP_006712334.1 | 32, 33 | 4716 | |||
GenBank mRNA | AB095944.1 | - | 3, 4 | 773 | ||
AK126998.1 | - | 10, 11 | 1392 | |||
AK127553.1 | - | 6, 7 | 669 | |||
AK128490.1 | - | 16, 17 | 2143 | |||
AK303062.1 | - | 7, 8 | 763 | |||
AL834144.1 | - | 4, 5 | 370 | |||
BX649036.1 | - | 8, 9 | 899 | |||
CR749469.1 | - | 10, 11 | 1165 |