Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2CR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127391.2 | NP_001120863.1 | 15 | 1654 | ||
NM_001289993.1 | NP_001276922.1 | 15 | 1322 | |||
NM_139163.3 | NP_631902.2 | 15 | 1723 | |||
NR_110620.1 | 14 | 1820 | ||||
XM_011510606.2 | XP_011508908.1 | 15 | 1669 | |||
XM_011510610.2 | XP_011508912.1 | 13 | 1114 | |||
XM_011510612.2 | XP_011508914.1 | 9 | 821 | |||
XM_017003360.1 | XP_016858849.1 | 14 | 1337 | |||
XM_017003362.1 | XP_016858851.1 | 13 | 1155 | |||
GenBank mRNA | AB053314.1 | - | 14 | 1308 | ||
AK124115.1 | - | 14 | 1766 | |||
AK292283.1 | - | 15 | 1322 | |||
BC031603.1 | - | 15 | 1634 | 5168160 | ||
BX095441.1 | - | 5 | 469 |